Spécifications
- Description:Cleaning Basket
- Supplier No.:62110
Spécifications
A propos de cet article
Made from stainless steel. Keeps small parts together when cleaning ultrasonically. A spring handle applies firm pressure to the basket to keep it closed when in use. Its overall length is 6". Spherical basket has a 38 mm (1 1/2") diameter.