Caps for Syringes, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip
These tips make the syringes suitable for usage as embedding media storage vessels.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.