EMS Zirconia Ceramic Tip Tweezers, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These tweezers have Zirconia ceramic precision tips.
- Light weight
- Aluminum handles
Surface resistively 10E12 ohm, Contamination free, high temperature (1000 °C). All styles have length of 134 mm (5¹/₄").
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.