One-Piece PFA Fittings with Integrated Ferrules, S.C.A.T.
# de catalogue 10040-250
Fournisseur: S.C.A.T.
One-Piece PFA Fittings with Integrated Ferrules, S.C.A.T.
# de catalogue 10040-250
Fournisseur: S.C.A.T.
Spécifications
- Description:3.2 mm ID, red
- Cat. no.:10040-250
- Supplier No.:107064
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.65 to $603.92
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Fournisseur: MicroSolv Technology Corp
$76.74
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$182.38 to $196.03
SureSTART™ Glass Crimp Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$1,951.02 to $1,951.02
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$131.41 to $136.31
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$415.75