SafetyWasteCaps with Mechanical Level Control, S.C.A.T.
Fournisseur: S.C.A.T.
Certificats
A propos de cet article
For safe disposal of fluid waste, with red level float indicating fluid level.
- Especially recommended for HPLC Systems
- Chemically resistant to aggressive organic solvents
Collect your solvent waste safely, without being exposed to hazardous gases and solvent vapours. The integrated exhaust filter absorbs 99% solvent vapours, thereby protecting staff and the working environment. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
Available for multiple screw threads and container sizes.
With safety funnel with automatic closure: Manually operated safety funnel which is only opened when disposing waste fluids. When releasing the opening mechanism, the container closes automatically.
Ordering information: Exhaust filter has to be ordered separately.
Customers Who Bought This Also Bought