Fire Resistant SafetyWasteCaps, S.C.A.T.
# de catalogue 10040-180
Fournisseur: S.C.A.T.
Spécifications
- Description:GL 45 screw caps, PP, For exhaust filter, 1x tubing 6,4 mm ID and 4x capillary connections 2.3 and 3.2 mm (3/32 and 1/8") OD
- Cat. no.:10040-180
- Supplier No.:112921
Spécifications
A propos de cet article
Keep your laboratory safe and protect your health and environment.
- Suitable for any HPLC system by multiple capillary and tubing connectors
- Only pure PTFE is used to guarantee maximum chemical resistance
- No unfiltered evaporation of hazardous gases
Customers Who Bought This Also Bought