Corning® 96-Well Black Clear Bottom Plates with Lid, Fibronectin, Nonsterile, Corning
Catalog # 10037-564
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Format:96-well plate
- Sterility:Non sterile
- Color:Black/Clear Bottom
- Description:96-Well Clear Bottom, with Lid
- Cat. no.:10037-564
- Surface Treatment:Fibronectin
- Supplier No.:4594
Specifications
About this item
These microplates are ideal for promoting cell attachment, proliferation, and differentiation of a variety of cell types.
Frequently Bought Together