SP Bel-Art Large Molded Fume Hood with Baffle, Bel-Art Products, a part of SP
# de catalogue 30181-797
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Fume Hood with Baffle
- Height:30"
- Cat. no.:30181-797
- Shipping Weight:56 lbs.
- Length:42"
- Width:20"
- Supplier No.:500000004
Spécifications
A propos de cet article
This fume hood includes a flat internal baffle across the rear of the shell that slopes forward, leaving space beneath for air flow to sweep the deck, and space above in order to efficiently remove airborne vapors.
- One-piece design
- Polyethylene plastic construction
- Clear acrylic sash slides on inside tracks
- Sash facilitates continuous airflow
The one-piece shell is molded of polyethylene plastic with a clear acrylic sash that slides on inside tracks. It can be set in an open, closed or intermediate position. The front opening is 88 x 36 cmH (34½ x 14³/₁₆"). When in the closed position, the sash remains open slightly to facilitate continuous air flow. Stack O.D. is 15 cm (6").
Ordering information: An exhaust system capable of drawing 350 cfm (free air) is recommended.
Customers Who Bought This Also Bought