SP Bel-Art The Glascribe® Scriber Pen, Bel-Art Products, a part of SP
# de catalogue 13196-058
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:The Glascribe® Pen
- Cat. no.:13196-058
- Supplier No.:441500000
Spécifications
A propos de cet article
The Glascribe® Pen, with retractable tungsten carbide tip, (Mohs Hardness = 9) easily inscribes on glass, ceramics, and plastic.
- Rectractable tip
- Easy writes on glass, ceramics, and plastic
- Included pocket clip
The pen has a pocket clip, so it is always handy when needed.
Customers Who Bought This Also Bought