SP Bel-Art Small Storage Bin, Bel-Art Products, a part of SP
# de catalogue 47750-968
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Color:White
- Description:Small storage bin
- Cat. no.:47750-968
- L×W×H:12.7 x 15.2 x 17.8 cm (5 x 6 x 7")
- Supplier No.:186690001
Spécifications
A propos de cet article
Made from white acrylic with a clear acrylic front panel for viewing contents, this dispenser keeps items such as alcohol pads, finger cots, microcentrifuge tubes and other small lab supplies organized and within easy reach.
- White acrylic plastic construction
- Easy to clean
- Transparent acrylic lid for easy visibility
The top is removable for filling and a hinged cover allows easy dispensing of items as needed.
Customers Who Bought This Also Bought