SP Bel-Art Resharpenable Rigid Scraper, Bel-Art Products, a part of SP
# de catalogue 13197-404
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:28.5 cm (11¼”)
- Cat. no.:13197-404
- Supplier No.:368200000
Spécifications
A propos de cet article
Made from glass-reinforced polyimide nylon, this rigid scraper's straight, chisel-type edge may be resharpened.
- Glass-reinforced polyimide nylon construction
- Rigid design
- Straight edge
- Wide, flat chisel-type blade
It has a 14 cm (5½”) long x 6.3 cm (2.5”) wide flat blade. FDA-grade materials are suitable for direct contact with foods, drugs, and cosmetics. Highest temp: 180ºF (82ºC).
Customers Who Bought This Also Bought