SP Bel-Art Scrapers, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Smooth, chemical-resistant plastic scrapers are useful for removing residues from filters, cleaning containers, mixing and distributing solids in fluids, spreading pastes over large areas, and many other scraping, spreading, stirring or residue removing tasks.
- Smooth, chemical-resistant surface
- Available in LDPE or HDPE models
- Used for a variety of applications
- Chisel edge on one end, hexagonal shank on the other
The blade is 12.7 cm (5”) wide x 15.2 cm (6”) long and tapers from 9.5 mm (⅜”) down to a chisel edge. The upper end extends into a hexagonal shank 35 mm (1⅜”) wide. Available in white, FDA-grade LDPE or red HDPE.
Customers Who Bought This Also Bought