SP Bel-Art Long Handled Scraper, Bel-Art Products, a part of SP
# de catalogue 57925-032
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:152 cm (60")
- Cat. no.:57925-032
- Supplier No.:368290001
Spécifications
A propos de cet article
Semi-Flexible nylon scraper attached to a rigid fiberglass handle provides a secure grip when reaching into deep vessels.
- Nylon scraper will not scratch glass
- Scraper, when removed from handle, can be sterilized at 100 °C
- Handle includes top opening for hanging
The scraper/paddle is 11.4×17.8 cm long (4.5×7"), with one rounded corner.
Certifications: Meets FDA Regulation Title 21 paragraphs 170-199, 21 CFR 177.1500 and EEC regulations 90/128EEC, 92/93EEC and 95/3/EC.
Customers Who Bought This Also Bought