SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
# de catalogue 70680-276
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:19.1 cm (7½”)
- Cat. no.:70680-276
- Supplier No.:378260000
Spécifications
A propos de cet article
A smooth, tapered, stainless sampler for extracting a “plug” of material from bags or blocks of cheese, grease, soap, clay, etc.
- Stainless steel construction
- Can easily cut through a bag or box
- Smooth, tapered design
The sharp end of the satin finish sampler will cut through a bag or box.
Twisting the handle one-half turn will cut out a plug of material 15.2 cm (6”) long tapering from 1.7 cm (¹¹/₁₆”) to 1.3 cm (½”). “T” handle is 7 cm (2¾”).
Customers Who Bought This Also Bought