PYREX® Capillary Melting Point Tubes, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Quality glassware at an affordable price.
- Disposable
- Available open-ended on both sides or closed on one side
- Made from no. 7740 glass
All tubes have a 0.2 to 0.25 mm wall range.
Customers Who Bought This Also Bought