Boekel Scientific Tight Temperature Incubators
Fournisseur: Boekel
Certificats
A propos de cet article
The Boekel TTT Incubator is specifically designed for maximum temperature stability and uniformity while using forced air convection for those temperature sensitive applications. Ideal for water/wastewate protocols or any microbiology and biology protocol requiring tight uniformity and temperature.
- Large viewing window
- Easy to use digital control system
- User configurable alarm and timer
- Stackable
- Removable shelf
- Designed for benchtops with limited space
Using mechanical convection and an optimized configuration it is ideal for any small sample temperature sensitive process.
Delivery: One removeable shelf is included with each unit.
Spécifications
- Catalog No:
- 10027-268
- 10027-266
- Shelves supplied (max.):
- 1
- 1
- Temperature range:
- Ambient +8.0…70 °C
- Ambient +8.0…70 °C
- Temperature uniformity:
- ±0.5 °C at 37 °C
- ±0.5 °C at 37 °C
- Exterior dimensions:
- 12.0W×6.0D×8.0"H
- 12.0W×6.0D×8.0"H