SP Bel-Art Stainless Steel Spoons, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These double-ended, 304 stainless steel sampling spoons have a solid, rounded shaft.
- Double-ended design
- Stainless steel construction
- Optional FEP coating provides non-stick surface
Available in plain polished stainless steel or coated with Fluo-Kem® Teflon® FEP, which complies with FDA regulations governing direct food contact applications.
Customers Who Bought This Also Bought