About this item
The VWR® Standard Universal Homogenizing Package is designed for use with all types of samples, even tough or stringy.
- Designed for use with all types of samples, even tough or stringy
- Allows flexibility
- Fast and efficient operation
The package allows the flexibility of homogenizing within a wide range of tube sizes, from micro-tubes to large tubes (1.5–50 mL tubes)
The VWR® Deluxe Universal Homogenizing Package is also designed for use with all types of samples, even tough or stringy. This package allows the flexibility to process tubes ranging from 1.5–50 mL as well. The 7mm and 10mm saw-tooth generator probes provide fast and efficient operation. From soft to tough samples, both saw-tooth generator probes provide extra cutting action during homogenizing, ensuring fast and complete sample breakdown. The package is supplied complete with stand assembly for hands-free homogenizing within sizes ranging from micro-tubes to large tubes, without having to hold the homogenizer unit.
Both the 7 mm and 10 mm saw-tooth generator probes provide fast and efficient operation. From soft to tough samples, both saw-tooth generator probes provide extra cutting action during homogenizing, ensuring fast and complete sample breakdown.
Specifications
- Catalog No:
- 10032-332
- 10032-334
- Weight:
- 0.64 kg (1.4 lbs.)
- 0.64 kg (1.4 lbs.)
- Dimensions:
- 5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")
- 5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")
- Power rating:
- 144 W
- 144 W
- Sound level:
- 68 dB
- 68 dB