Photometer NOVA 30 A Spectroquant® | Avantor
Photometer NOVA 30 A Spectroquant®
Photometer NOVA 30 A Spectroquant®
Supplier:  
Certificates
0 Options Available Below
Product Details & Documents
The Spectroquant® NOVA 30A is capable of running all the important cell tests for wastewater analysis.
  • Barcode reading of all Spectroquant® tests: Automatic selection of methods with presentation of the results
  • Integrated QA/QC checks: Three modes, checking instrument, method and sample volume
  • Turbidity correction
Suitable for some critical tests, measurement to ppb levels, such as cyanide.
Specifications for Product Family
Spécifications
  • Catalog No:
  • EM1.09748.0001
  • CA1.09748.0001
  • Model:
  • NOVA 30A
  • NOVA 30A
  • Measuring modes:
  • Absorbance, transmittance, concentration
  • Absorbance, transmittance, concentration
  • Photometric range:
  • –0.300 to 3.200 A
  • –0.300 to 3.200 A
  • Photometric reproducibility:
  • 0.001 - 1.000 A
  • 0.001 - 1.000 A
  • Photometric resolution:
  • 0.001 A
  • 0.001 A
  • Wavelength:
  • 6 filters in array technique, 340, 445, 525, 550, 605 690 nm
  • 6 filters in array technique, 340, 445, 525, 550, 605 690 nm
  • Cells:
  • 16 mm diameter
  • 16 mm diameter
  • Interfaces:
  • RS232
  • RS232
  • Light source:
  • Tungsten halogen lamp
  • Tungsten halogen lamp
  • Method storage:
  • 60 methods for Spectroquant® cell tests
  • 60 methods for Spectroquant® cell tests
  • Power requirements:
  • 100 - 240 V
  • 100 - 240 V
  • W×D×H:
  • 260×270×140
  • 260×270×140
  • Weight:
  • 2.8 kg
  • 2.8 kg
  • Storage capacity:
  • 500 rsults
  • 500 results
Spécifications
Product Family Options
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Packaging
    Availability
    Price
  • Halogen Lamp
    Halogen Lamp
    CA1.09749.0001
    NOVA 30/60 Spectroquant®
    Plastic bag
    Log in pour la disponibilité
    Each - $262.54