Germicidal Ultraviolet Lamp, Labconco
Catalog # 62403-141
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Cat. no.:62403-141
- Voltage:115 V
- Power:30 W
- Weight:5 lbs.
- Supplier No.:1271300
Specifications
About this item
This germicidal UV lamp is for use in Labconco PuriCare Procedure Stations (pre-2008), Purifier Cell Logic Biosafety Cabinets, Purifier Delta Class II Biosafety Cabinets, and Purifier Logic Class II Biosafety Cabinets.
Customers Who Bought This Also Bought