Dura-Grind™ Stainless Steel Dounce Tissue Grinder, Wheaton®
Fournisseur: Avantor
Certificats
A propos de cet article
Wheaton Dura-Grind™ stainless steel Dounce tissue grinders provide a rugged alternative to glass Dounce tissue grinders.
- Smooth pestle action requires less effort than glass tissue grinders
- Precision machined from number 316 stainless steel
- Mortar has a flat bottom allowing it to stand upright
- Unit cannot be autoclaved
Single pestle and mortar as a matched set to exacting tolerance of 0.013 mm.
Customers Who Bought This Also Bought