VWR® UV-3100PC, V-3000-PC Spectrophotometers, UV-Vis Scanning and Vis
Supplier: VWR International
Certificates
About this item
Two fully scanning spectrophotometers for advanced measurements and quality control.
- Large LCD display (320×240 pixels)
- Full scan mode
- Easily accessible USB port for data import and export
- Supplied application software 'UV-Vis Analyst' allows PC control of the spectrophotometer
- Methods for PC control or standalone include: Basic mode, quantitative, wavelength scan, kinetics, multi-wavelength, DNA/protein
- The range of accessories includes an automatic 4-cell changer and Peltier thermostatted single cell holder
Reliable, robust and easy to use instruments with a range of accessories that will support the creation of a personal spectrophotometer measuring station. The application software offers all the important methods expected from a 'universal’ instrument. All housed in a robust cover with a well designed optical bench.
Ordering information: Supplied with 4 optical glass cells, 2 quartz cells (excludes model V3000-PC), dust cover, USB cable, USB (8 GB) stick, and UV-Vis analyst software for PC control.
Specifications
- Catalog No:
- 10037-438
- Display:
- 5” graphic LCD (320×240 pixels)
- Optical design:
- Single beam, grating 1200 lines/mm, silicon photodiode detector
- Photometric accuracy:
- ≤±0.5% T or 0.005 A at 1 A
- Photometric range:
- −0.3 to 3 A; 0 - 200% T; 0 - 9999 Conc
- Photometric reproducibility:
- ±0.2% T
- Photometric stability:
- ±0.002 A/h at 500 nm
- Wavelength accuracy:
- ±0,5 nm
- Wavelength repeatability:
- ≤0,2
- Electrical:
- 120V
- Interfaces:
- USB port to PC / parallel port to printer
- Lamp source, lifetime:
- Tungsten/deuterium
- Languages:
- EN, DE, ES, FR
- Light source:
- Tungsten/deuterium
- No. of standards:
- 200 standard curves
- Scan speed:
- Low, medium, high (max. 3000 nm/min)
- Storage capacity:
- 200