VWR® Double Beam UV-Vis Spectrophotometer
10037-442
- Description:UV-6300PC Spectrophotometer
- Cat. no.:10037-442
A double beam spectrophotometer suitable for a wide range of applications including general research, pharmaceutical, biochemical and clinical laboratory use, new material development, and quality control.
- Standard photometrics - absorption, transmission, concentration
- Quantitative analysis and kinetics
- Multi-wavelength scan, spectrum scan
- DNA/RNA and protein analysis
The instrument can be used in stand-alone mode or in conjunction with a PC using the supplied UV-VIS Analyst software which then delivers more powerful data processing, expanded data collecting, and removes the limit on storage capability.
: Supplied with four optical glass cells, two quartz cells, dust cover, USB cable, USB (8 GB) stick, UV-Vis analyst software for PC control and single cell holders for sample and reference.