Torque-Rite® Nuts, Rubber Fab
Fournisseur: Rubber Fab
Certificats
A propos de cet article
Torque-Rite® allows you to control compression and expansion while maintaining constant inch/pounds force insuring a perfectsSurface I.D
when used with a Rubber Fab sanitary gasket. Torque-Rite® eliminates the problems associated with over - or under-tightening a gasket which can lead to an unsanitary system.
Customers Who Bought This Also Bought