ESD Knit Hip-Length Lab Coats
Fournisseur: VWR International
Certificats
A propos de cet article
The ESD Knit Lab Coat is designed to provide excellent anti-static protection for electronics applications.
- Excellent static dissipative properties
- Lightweight, breathable fabric for added comfort when worn over regular clothes
- Reinforced front facing and flat felled seams for improved durability
- Three-position snap cuff for adjustability
The garments feature two lower pockets and one breast pocket. Each lab coat features a snap-front with reinforced facing and flat-felled seams for extra durability, and a three-position snap cuff.
Manufactured with a durable and breathable fabric, it offers a slight stretch for added comfort and ease of movement during asembly operations.
Ideal for use in micro-electronics assembly, microchip manufacturing, and semi-conductor production.
Ordering information: Available in blue only. Sizes 3X-Large and 4X-Large require extended lead time. Contact your Avantor sales representative for more information.
Spécifications
- Catalog No:
- 10032-394
- 10032-314
- 10032-768
- 10032-770
- 10032-310
- 10032-312
- 10032-308
- Garment application:
- Electronics/semi-conductor
- Electronics/semi-conductor
- Electronics/semi-conductor
- Electronics/semi-conductor
- Electronics/semi-conductor
- Electronics/semi-conductor
- Electronics/semi-conductor
- Risk area:
- Anti-static, ESD
- Anti-static, ESD
- Anti-static, ESD
- Anti-static, ESD
- Anti-static, ESD
- Anti-static, ESD
- Anti-static, ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- 94% polyester, 6% carbon
- Weight:
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)
- 2.21 oz./sq. yd. (±5%)