A propos de cet article
This scrub top provides an excellent base-layer of protection for any cleanroom garment system.
- Pullover design with three-snap Henley-style collar that provides added room for easy on/off
- Binding on collar helps maintain shape and uniformity
- Raglan-style sleeves gives end-user increased mobility
- Sleeves include color-coordinated filament knit cuff for extra comfort
- Color-coded loops for easy size identification
- 5" side slits at bottom of garment for added comfort and long-term durability
Manufactured with a durable and lightweight micro-denier continuous filament polyester material to elimate a source for lint. The scrub tops also feature a sandwash finish for soft-touch comfort, and moisture wicking properties.
It has a pullover design with a three-snap Henley collar, raglan sleeves, and 12.7 cm (5") side slits for added mobility. Sleeves include filament knit cuffs for extra comfort under cleanroom coveralls and gloves.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10032-402
- 10032-404
- 10032-398
- 10032-400
- 10032-396
- 10032-408
- 10032-410
- 10032-406
- Garment application:
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- 100% polyester sandwash fabric
- Weight:
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.