A propos de cet article
These scrub pants provide an excellent base-layer of protection for any cleanroom garment system
- Drawstring front/elastic back
- Double needle felled seams
- Color-coded sizing loop
- Lint-free fabric
Manufactured with a durable, lightweight micro-denier continuous filament polyester material to elimate a source for lint. The material has a sandwash finish for soft-touch comfort and moisture wicking properties.
They feature a combination elastic back and drawstring front for secure comfort, and the leg opening has an elastic back with a three-position snap for an adjustable closure.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10032-424
- 10032-426
- 10032-420
- 10032-422
- 10032-416
- 10032-418
- 10032-412
- 10032-414
- Garment application:
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Medical, Food, Cleanroom, General Purpose, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- 100% polyester, sandwash fabric
- Weight:
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.
- 2.6 oz./sq. yd.