Stirring Shaft, Borosilicate, Ace Glass Incorporated
# de catalogue 89075-074
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Length:44 cm
- Description:Hollow Borosilicate Glass Stirring Shaft
- Cat. no.:89075-074
- Supplier No.:9532-10
Spécifications
A propos de cet article
Precision ground glass stirring shaft with attached vanes, 10 mm
For use when agitation and gas introduction are desired at the same time. Agitator will fit through a standard taper 45/50 joint. Designed for use with ACE Trubore® bearing and mini-lab flask heads. Introduced gas exits the hollow shaft within the agitator and produces a fine dispersion. Join to stirrer motor with series chucks.
Customers Who Bought This Also Bought