Micro-Hematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These Micro-Hematocrit Capillary Tubes are manufactured from soda lime glass, packed in plastic vials with reclosable snap-lock caps to maintain cleanliness.
Ends may be sealed by flame, sealing clay or patented caps. Heparinized capillary tubes are treated to prevent blood clotting.
Customers Who Bought This Also Bought