Samco™ Special Purpose Transfer Pipettes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Samco™ Special Purpose Transfer Pipettes are ideal for delivering drops onto slides.
Made of pliable, nontoxic polyethylene, the low-affinity surface reduces loss of cells and valuable proteins due to binding.
Customers Who Bought This Also Bought