ContactUsLinkComponent
Environmental and Stability Test Chambers, Extra Large Capacity, Caron Products
Environmental and Stability Test Chambers, Extra Large Capacity, Caron Products
Fournisseur:  Caron Products
Certificats
Environmental and Stability Test Chambers, Extra Large Capacity, Caron Products
Environmental and Stability Test Chambers, Extra Large Capacity, Caron Products
Fournisseur:  Caron Products
Certificats
Spécifications
  • Catalog No:
  • 10033-898
  • 10033-836
  • 10033-940
  • 10033-938
  • 10033-902
  • 10033-900
  • Temperature range:
  • 5…70 °C (41…158 °F)
  • 5…70 °C (41…158 °F)
  • 5…70 °C (41…158 °F)
  • 5…70 °C (41…158 °F)
  • 5…70 °C (41…158 °F)
  • 5…70 °C (41…158 °F)
  • Temperature control:
  • ±0.1°C
  • ±0.1°C
  • ±0.1°C
  • ±0.1°C
  • ±0.1°C
  • ±0.1°C
  • Humidity range:
  • 20 to 95%
  • 20 to 95%
  • 20 to 95%
  • 20 to 95%
  • 20 to 95%
  • 20 to 95%
  • Finish:
  • Stainless Steel Interior, Stainless Steel Exterior
  • Stainless Steel Interior, Stainless Steel Exterior
  • Stainless Steel Interior, Stainless Steel Exterior
  • Stainless Steel Interior, Stainless Steel Exterior
  • Stainless Steel Interior, Stainless Steel Exterior
  • Stainless Steel Interior, Stainless Steel Exterior
Spécifications
Informations connexes
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product.
Extended Warranty Packages offer peace of mind with up to an additional 1 year or 3 years on top of the original warranty, covering parts and labor for any manufacturer's issues that may occur during this time. Purchase of an Extended Warranty is only valid prior to the original warranty expiration and is only available for U.S. Customers. See table below for more information.
PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
PM Service Kits for Accessories help to minimize downtime by providing routine maintenance items for accessories. See table below for more information.