Reaction Head for Pilot Plant, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
KF Flange Flat Borosilicate Glass Head with holes to accept PTFE standard taper joint inserts.
PTFE Inserts:
89427-918 24/40 PTFE Insert
89427-920 29/42 PTFE Insert
89427-922 45/50 PTFE Insert
89427-924 71/60 PTFE Insert
Customers Who Bought This Also Bought