Ultrasonic Footswitch, Ace Glass Incorporated
# de catalogue 89075-518
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Ultrasonic Footswitch, for 600 and 1500 W Power Supplies
- Cat. no.:89075-518
- Supplier No.:9824-43
Spécifications
A propos de cet article
For use with 600W or 1500W Power Supply, to enable hands-off operation
Customers Who Bought This Also Bought