Ultrasonic Booster, Ace Glass Incorporated
# de catalogue 89075-516
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Ultrasonic Booster
- Cat. no.:89075-516
Spécifications
A propos de cet article
When connected between the converter and horn, the booster increases the amplitude of vibration at the horn tip by a factor of two
Use to process very difficult applications
Customers Who Bought This Also Bought