SP Bel-Art Adjustable Microplate Tilting Stand, Bel-Art Products, a part of SP
# de catalogue 62866-107
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Cat. no.:62866-107
- Supplier No.:189380000
Spécifications
A propos de cet article
Make precise additions of liquid components to microplate wells by adjusting plates to the most convenient angle.
- Epoxy coating
- Clear acrylic platform
- Durable steel base
- Fully adjustable for providing convenient microplate angles
- Compatible with all sizes of microplates
The Tilting Stand helps reduce errors in non-manifold additions of assay components to a multiwell microplate, including expensive, hazardous, or radioactive materials
An epoxy-coated steel base provides stability, and the hand-adjustable clamping screws maintain the desired angle. The stand holds all sizes of microplates. Maximum tilting angle is 45°. Pivots on 2 screws.
The angular presentation of the plate and clear acrylic platform beneath allows users to easily judge which wells have received liquid addition by comparing the menisci between neighboring wells.
Customers Who Bought This Also Bought