ContactUsLinkComponent
Button Type Stirring Shaft, 5 mm, Hollow Glass, Ace Glass Incorporated
Button Type Stirring Shaft, 5 mm, Hollow Glass, Ace Glass Incorporated
# de catalogue 89075-084
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
9535 STIRRING SHAFT, 5mm
Button Type Stirring Shaft, 5 mm, Hollow Glass, Ace Glass Incorporated
# de catalogue 89075-084
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Hollow Glass Stirring Shaft
  • Cat. no.:
    89075-084
  • Shaft Diameter:
    5 mm
  • Supplier No.:
    9535-06
Spécifications
A propos de cet article
Precision ground, hollow, 5 mm glass stirring shaft
Button type shaft accepts blades and can be used in matching bearings. Holes are provided in the side of the shaft for material introduction and at the bottom for emission into the flask contents. Join to stirrer motor using certain Ace series chucks.
Customers Who Bought This Also Bought