ContactUsLinkComponent
Precision Ground Stirring Shaft, Ace Glass
Precision Ground Stirring Shaft, Ace Glass
# de catalogue 89075-076
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
9533 STIRRING SHAFT, Glass, 10mm, Precision Ground
Precision Ground Stirring Shaft, Ace Glass
# de catalogue 89075-076
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Borosilicate Glass Stirring Shaft, Precision Ground, 10 mm
  • Cat. no.:
    89075-076
  • Supplier No.:
    9533-02
Spécifications
A propos de cet article
Precision ground glass stirring shaft, 10 mm O.D., attached vanes
Vanes will fit through a standard taper 45/50 joint and are contoured for use with Ace series Mini-Lab flasks. Use with Trubore® bearings. Compatible with Ace mechanical seal and PTFE collet type bearings. Join to stirrer motor using series chucks.
Customers Who Bought This Also Bought