Complete Photochemical Reaction System, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Complete reaction assembly with all parts needed for immediate operation
Borosilicate glass reactor has a standard taper 60/40 center joint, one- stadnard taper 14/20 angled joint for sparger tube, one-standard taper 24/40 vertical joint for condenser, and one-#7 Ace-Thred joint to accommodate thermometer. Volumes indicated are total volumes. Volume in reactive area of lamp is 40-50% of the total volume. Complete assembly consists of reactor, quartz immersion well, sparger tube, PTFE stir bar #7 thermometer bushing, three meters of 4.8 mm (3/16") I.D. PTFE tubing, stand, 450 watt lamp and 450 watt power supply.
Customers Who Bought This Also Bought