Photochemical Reaction Vessel, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Reaction vessel constructed of borosilicate glass to accommodate the immersion wells
With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one- #7 Ace-Thred side arm for thermometer. Center joint is standard taper 60/40. Bottom of reactor is flat to allow use of magnetic stirrer. Volumes indicated are total volumes. Volume in reactive area of lamp is approx. 40-50% of total volume.
Ordering information: Complete unit consists of vessel, sparger tube, PTFE stir bar, threaded nylon bushing and three meters of 4.8 mm (3/16") I.D. PTFE tubing.
Customers Who Bought This Also Bought