Photochemical Jacketed Reaction Vessel with Stopcock Bottom, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Jacketed reaction vessel to enable cooling of reactant material during photolysis plus the added feature of a 2 mm bore, 1:5 PTFE stopcock at bottom for draining inner vessel contents
Constructed of borosilicate glass to accommodate immersion wells. With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one-#7 Ace-Thred side arm for thermometer. Center joint is standard taper 60/40. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40-50% of total volume. Size C hose connections. Listing is vessel only.
Customers Who Bought This Also Bought