Drying Tubes with Rubber Stopper, Ace Glass Incorporated
# de catalogue 89074-486
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- ST Joint No.:14/20
- Description:Drying Tubes
- Cat. no.:89074-486
- Overall Length:180 mm
- Supplier No.:9419-03
Spécifications
A propos de cet article
This drying tube adapter has an inner standard taper joint, and comes complete with rubber stopper and ¼×2" tube.
Fabricated from borosilicate glass.
Ordering information: Stopper/tube can be ordered separately.
Customers Who Bought This Also Bought