SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
# de catalogue 89087-688
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Foil cover fits 1.75" wall height, 2" base diameter; 5.5" diameter
- Cat. no.:89087-688
- Supplier No.:F132001055
Spécifications
A propos de cet article
Used in cell culture to cover flasks while allowing proper oxygenation, these economical and disposable covers also provide dust-free storage of clean labware.
- Provides a temporary, sterile boundary for autoclave sterilization of labware
- Preformed to quickly cover a wide variety of labware using only one hand
Great for use in centralized washing areas where cleaned labware needs to be transported back to the lab while remaining clean and contaminant-free.
Customers Who Bought This Also Bought