Photochemical Quartz Double-Walled Immersion Well, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Double-walled, quartz immersion well for use with certain U.V
- Well has a standard taper 60/40 inner joint
- Length above joint is 76 mm
Ace lamps. With inlet and outlet tubes for cooling. A small diameter inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet
Customers Who Bought This Also Bought