Photochemical Safety Reaction Cabinets, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This steel cabinet allows for the safe operation of ACE photochemical reaction equipment
Inside the cabinet is a plug-in light, auxiliary 120 V socket and a 60 CFM exhaust fan, all controlled by an on/off switch. Also inside are pin jack sockets for lamp connection and a removable 1/2" aluminum rod, mounted vertically, for clamping the reactor.
Eliminates the need for a hood, or to construct a special safe area to operate the U.V. lamp. The cabinet has welded seams and a fully hinged door with lip to prevent light from escaping. The door has a key lock for positive closure and it controls a safety switch that prevents U.V. lamp operation unless door is closed. The floor of cabinet is sealed to one-inch height to contain any possible spills.
Ordering information: The cabinet is supplied with a six-foot grounded power cord with NEMA plug for connection to a 120 V source, and a six-foot, two-wire cord with male pin jacks for connection to the lamp power supply. Handles are mounted on both sides for easier carrying, and there are rubber feet on the bottom of the cabinet for stability. Measures: 36" high x 211/4" wide x 181/2" diameter. Painted black inside, chemically resistant blue outside. Weighs 60 lbs.
Customers Who Bought This Also Bought