Ultrasonic Reactor, Small Volume, Ace Glass Incorporated
# de catalogue 89075-524
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Compete Reaction Assembly
- Cat. no.:89075-524
- Supplier No.:9830-25
Spécifications
A propos de cet article
Complete reaction assembly with parts necessary to perform mixing and reactions from 3 to 200 mL
Includes three borosilicate glass vessels, power supply with converter, 1/2" horn, 1/2" extender, 1/4" microtip, slide adapter and clamp.
Customers Who Bought This Also Bought