Photochemical Borosilicate Glass Double-Walled Immersion Well, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Double-walled, borosilicate glass immersion well for use with U.V
- Well has a standard taper 60/40 inner joint
- Outer jacket is 53 mm O.D.
- Length above joint is 76 mm
Optional PTFE tube attached to a glass tube and secured wioth a #7 Ace-Thred bushing. PTFE tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet.
lamps. With inlet and outlet tubes for cooling. A small diameter inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet.
Customers Who Bought This Also Bought