SP Bel-Art Weighted Beaker/Flask Holders with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Weighted holder grips beakers or flasks firmly with four flared stainless steel prongs.
- Stainless steel prongs
- Weight is made of vinyl-coated lead
- Compatible for use in a boiling liquid
- Low center of gravity
The Vikem® vinyl-coated lead ring weight secured beneath the holder keeps container immersed in water, provides a low center of gravity and allows water to circulate underneath the beaker. Can be used in boiling liquid up to 135˚C(275˚F) for 1 hour.
Frequently Bought Together