SP Bel-Art Filtering Flask with Side Arm, Bel-Art Products, a part of SP
# de catalogue 47750-576
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Capacity:1000 ml
- Cat. no.:47750-576
- Supplier No.:389410000
Spécifications
A propos de cet article
Polypropylene plastic flask has side-arm at 45˚ downward angle to prevent tipping.
- Polypropylene construction
- Downward angle prevents tiping
- Flexible side arm
Arm accepts flexible tubing from ⅜" to ½" I.D.
Customers Who Bought This Also Bought