Ultrasonic Horn Extender, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Titanium extender screws into threaded end of ultrasonic horn
This accessory lengthens the horn (probe) by 5" or 10" for more versatility. Extenders have solid ends. 1/2" extender has single groove on 5-inch size and two grooves on 10" size for use with #15 Ace-Thred. 3/4" and 1" extenders do not have grooves. Order extender diameter to match horn diameter.
Customers Who Bought This Also Bought