Photochemical Immersion Well Absorption Sleeves, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Filter sleeve for use with all ACE Immersion Wells to restrict portions of the radiated energy from reaching the reactant material
An invaluable aid in predetermining which portion of the spectrum creates the reaction. Sleeves are glass, open end tubes which telescope into well assembly to surround the light source. For use with 100, 200, 450, and 1200 watt lamps, only.
Customers Who Bought This Also Bought